Learn More
Sino Biological 4E-BP1 / EIF4EBP1 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10022RP01400
Produced in rabbits immunized with purified, recombinant Human 4E-BP1 / EIF4EBP1 (rh EIF4EBP1; Catalog#10022-H07E; NP_004086.1; Ser2-Ile118). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0683