Learn More
Sino Biological 4E-BP1 / EIF4EBP1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10022T1650
Produced in rabbits immunized with purified, recombinant Human 4E-BP1 / EIF4EBP1 (rh 4E-BP1 / EIF4EBP1; Catalog#10022-H07E; NP_004086.1; Ser2-Ile118). 4E-BP1 / EIF4EBP1 specific IgG was purified by Human 4E-BP1 / EIF4EBP1 affinity chromatography.
Catalog No. 50-161-0686