Learn More
Sino Biological Nestin Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100244T08200
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human Nestin., and purified by antigen affinity chromatography.
Catalog No. 50-161-0704