Learn More
Sino Biological GJA1 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 100246R105100
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human GJA1. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0712