Learn More
Sino Biological Chromogranin A Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 100256T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human Chromogranin A., and purified by antigen affinity chromatography.
Catalog No. 50-161-0735