Learn More
Sino Biological Chromogranin A Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100256T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human Chromogranin A, and purified by antigen affinity chromatography.
Catalog No. 50-161-0740