Learn More
Sino Biological GSTπ (GlutathioneS-Transferase,pi) Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100259T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human GSTπ(GlutathioneS-Transferase,pi), and purified by antigen affinity chromatography.
Catalog No. 50-161-0747