Learn More
Sino Biological p63 (ΔN) Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100263T08200
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human p63(delta P63 /ΔN P63), and purified by antigen affinity chromatography.
Catalog No. 50-161-0756