Learn More
Sino Biological Androgenreceptor / AR Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100273T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Androgenreceptor / AR, and purified by antigen affinity chromatography.
Catalog No. 50-161-0771