Learn More
Sino Biological p63 / TP63 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100275T32100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human p63 / TP63, and purified by antigen affinity chromatography.
Catalog No. 50-161-0775