Learn More
Sino Biological Human Vimentin / VIM Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10028H08B50
A DNA sequence encoding the human VIM (Met 1-Glu466) (P08670) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-3840