Learn More
Sino Biological Notch1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100296T08100
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human Notch1 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0787