Learn More
Sino Biological Desmin Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 100301MM18200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the C-terminus of the Human Desmin. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0805