Learn More
Sino Biological Cytokeratin 19 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100307T36200
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the C-terminus of the human Cytokeratin 19. Human Cytokeratin 19 specific IgG was purified by Human Cytokeratin 19 affinity chromatography.
Catalog No. 50-161-0828