Learn More
Sino Biological KRT7 / Cytokeratin-7 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100310T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human KRT7 / Cytokeratin-7, and purified by antigen affinity chromatography.
Catalog No. 50-161-0854