Learn More
Sino Biological Cytokeratin 5 / KRT5 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100312T3850
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Cytokeratin 5 / KRT5, and purified by antigen affinity chromatography.
Catalog No. 50-161-0871