Learn More
Sino Biological GCDFP-15 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100314T0850
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human GCDFP-15 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0879