Learn More
Sino Biological IL9 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100318T40200
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the mouse IL9 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0887