Learn More
Sino Biological IL15 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100322T3250
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the mouse mIL15 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0897