Learn More
Sino Biological IL22 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100326T08200
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the mouse IL22., and purified by antigen affinity chromatography.
Catalog No. 50-161-0899