Learn More
Sino Biological Noggin / NOG Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100339T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Noggin/NOG, and purified by antigen affinity chromatography.
Catalog No. 50-161-0906