Learn More
Sino Biological CD45/PTPRC Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 100342R10150
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the C-terminus of the Human CD45/PTPRC.
Catalog No. 50-161-0920