Learn More
Sino Biological ARID3B Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100355T3250
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human ARID3B , and purified by antigen affinity chromatography.
Catalog No. 50-161-0952