Learn More
Sino Biological ALPL Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100356T08100
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human ALPL , and purified by antigen affinity chromatography.
Catalog No. 50-161-0959