Learn More
Sino Biological SNX6 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100358T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SNX6, and purified by antigen affinity chromatography.
Catalog No. 50-161-0969