Learn More
Sino Biological UCP1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100366T32200
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the C-terminus of the human UCP1 . Human UCP1 specific IgG was purified by Human UCP1 affinity chromatography.
Catalog No. 50-161-0984