Learn More
Sino Biological EGFR / HER1 / ErbB1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100372T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human EGFR / HER1 / ErbB1, and purified by antigen affinity chromatography.
Catalog No. 50-161-0987