Learn More
Sino Biological Anti-V5 Tag Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100378T365
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the V5-tag sequence. V5-tag specific IgG was purified by V5-taq affinity chromatography.
Catalog No. 50-161-1000