Learn More
Sino Biological Human CD27 / TNFRSF7 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10039H03H100
A DNA sequence encoding the extracellular domain (Met 1-Ile 192) of human CD27 (NP_001233.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the carboxy-terminus.
Catalog No. 50-161-3843