Learn More
Sino Biological Human CD27 / TNFRSF7 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10039H08B1200
A DNA sequence encoding the human CD27(P26842) ( Met 1-Ile192) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3846