Learn More
Sino Biological CD27 / TNFRSF7 Antibody (APC), Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10039R025A100
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD27 (rh CD27; Catalog#10039-H03H; Met 1-Ile 192; NP_001233.1) and conjugated with APC under optimum conditions, the unreacted APC was removed .
Catalog No. 50-161-1066