Learn More
Sino Biological CD27 / TNFRSF7 Antibody (FITC), Rabbit MAb 100Tests
Supplier:  Sino Biological 10039R025F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD27 (rh CD27; Catalog#10039-H03H; Met 1-Ile 192; NP_001233.1) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-1070