Learn More
Sino Biological CD27 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10039RP01400
Produced in rabbits immunized with purified, recombinant Human CD27 (rh CD27; Catalog#10039-H31H; P26842; Met1-Ile192). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-1080