Learn More
Sino Biological CD10 / Neprilysin / MME Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100390T0250
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CD10 / Neprilysin / MME, and purified by antigen affinity chromatography.
Catalog No. 50-161-1013