Learn More
Sino Biological Glypican 3 / GPC3 / OCI-5 Antibody (FITC), Rabbit MAb DFS Item
Manufacturer:  Sino Biological 100393R024F100
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Glypican 3 / GPC3 / OCI-5, and it was conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-1024