Learn More
Sino Biological LFA-1 / CD11a / ITGAL / Integrin alpha L Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100394T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human LFA-1 / CD11a / ITGAL / Integrin alpha L, and purified by antigen affinity chromatography.
Catalog No. 50-161-1034