Learn More
Sino Biological Human CD153 / CD30L / TNFSF8 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10040H01H5
A DNA sequence encoding the extracellular domain (Gln 63-Asp 234) of human CD30 Ligand (NP_001235.1) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-3851