Learn More
Sino Biological ADAM17 / CD156b Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100403T04200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human CD156b, and purified by antigen affinity chromatography.
Catalog No. 50-161-1085