Learn More
Sino Biological CXCR3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100409T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CXCR3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1100