Learn More
Sino Biological Human CD137 / 4-1BB / TNFRSF9 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10041H03H100
A DNA sequence encoding the N-terminal fragment (Met 1-Gln 186) of the extracellular domain of human 4-1BB (NP_001552.2) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3855