Learn More
Sino Biological Human CD137 / 4-1BB / TNFRSF9 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10041H08H50
A DNA sequence encoding the human 4-1BB extracellular domain (NP_001552.2) (Met 1-Gln 186) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3859