Learn More
Sino Biological CD137 / 4-1BB Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10041T16100
Produced in rabbits immunized with purified, recombinant Human CD137 / 4-1BB (rh CD137 / 4-1BB; Catalog#10041-H08H; NP_001552.2; Met1-Gln186). CD137 / 4-1BB specific IgG was purified by Human CD137 / 4-1BB affinity chromatography.
Catalog No. 50-161-1136