Learn More
Sino Biological PDX1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100419T36100
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human PDX1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1127