Learn More
Sino Biological Human 2B4 / SLAMF / CD244 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10042H08H100
A DNA sequence encoding the extracellular domain (Met 1-Arg 221) of human 2B4 (NP_057466.1) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3863