Learn More
Sino Biological 2B4 / CD244 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10042R0251
This antibody was obtained from a rabbit immunized with purified, recombinant Human 2B4 / CD244 (rh 2B4 / CD244; Catalog#10042-H08H; NP_057466.1; Met1-Arg221).
Catalog No. 50-161-1161