Learn More
Sino Biological 2B4 / SLAMF4 / CD244 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10042RP01100
Produced in rabbits immunized with purified, recombinant Human 2B4 (rh 2B4; Catalog#10042-H08H; NP_057466.1; Met 1-Arg 221). Total IgG was purified by Protein A affinity chromatography .
Catalog No. 50-161-1167