Learn More
Sino Biological Smad2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100425T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Smad2, and purified by antigen affinity chromatography.
Catalog No. 50-161-1148