Learn More
Sino Biological VEGFR2/KDR/Flk-1/CD309 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100429T10200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human VEGFR2/KDR/Flk-1/CD309, and purified by antigen affinity chromatography.
Catalog No. 50-161-1154