Learn More
Sino Biological HNF-1-alpha / HNF1A Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100434T02200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HNF-1-alpha / HNF1A, and purified by antigen affinity chromatography.
Catalog No. 50-161-1180