Learn More
Sino Biological NEFH Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100446T0850
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human NEFH, and purified by antigen affinity chromatography.
Catalog No. 50-161-1205